missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-iodopyrimidine, 98%

Manufacturer:  Alfa Aesar™ H6418503

 View more versions of this product

Catalog No. AAH6418503



Description

Description

Applications
Useful Intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store in dark. Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

5-Chloro-2-iodopyrimidine
White
874676-81-0
MFCD08062390
pyrimidine, 5-chloro-2-iodo, pubchem14957, acmc-209qnq, 5-chloro-2-iodo pyrimidine, ksc493q5d
C1=C(C=NC(=N1)I)Cl
240.428
240.43
98%
Light Sensitive
1g
C4H2ClIN2
14516398
BWDLBDLGVMLSCS-UHFFFAOYSA-N
5-chloro-2-iodopyrimidine
29938005
Powder

Chemical Identifiers

874676-81-0
240.428
BWDLBDLGVMLSCS-UHFFFAOYSA-N
29938005
C1=C(C=NC(=N1)I)Cl
C4H2ClIN2
MFCD08062390
pyrimidine, 5-chloro-2-iodo, pubchem14957, acmc-209qnq, 5-chloro-2-iodo pyrimidine, ksc493q5d
5-chloro-2-iodopyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

SDS Documents