missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-iodopyrimidine, 98%

$28.84 - $337.73

Chemical Identifiers

CAS 874676-81-0
Molecular Formula C4H2ClIN2
Molecular Weight (g/mol) 240.428
MDL Number MFCD08062390
InChI Key BWDLBDLGVMLSCS-UHFFFAOYSA-N
Synonym pyrimidine, 5-chloro-2-iodo, pubchem14957, acmc-209qnq, 5-chloro-2-iodo pyrimidine, ksc493q5d
PubChem CID 29938005
IUPAC Name 5-chloro-2-iodopyrimidine
SMILES C1=C(C=NC(=N1)I)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64185MD View Documents Alfa Aesar™
H64185MD
250mg Each for $28.84
 
AAH6418503 View Documents Alfa Aesar™
H6418503
1g Each for $85.14
 
AAH6418506 View Documents Alfa Aesar™
H6418506
5g Each for $337.73
 
Description

Description

Applications
Useful Intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store in dark. Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

874676-81-0
240.428
BWDLBDLGVMLSCS-UHFFFAOYSA-N
29938005
C1=C(C=NC(=N1)I)Cl
C4H2ClIN2
MFCD08062390
pyrimidine, 5-chloro-2-iodo, pubchem14957, acmc-209qnq, 5-chloro-2-iodo pyrimidine, ksc493q5d
5-chloro-2-iodopyrimidine

Specifications

Light Sensitive
250mg
C4H2ClIN2
14516398
BWDLBDLGVMLSCS-UHFFFAOYSA-N
5-chloro-2-iodopyrimidine
29938005
Powder
5-Chloro-2-iodopyrimidine
White
874676-81-0
MFCD08062390
pyrimidine, 5-chloro-2-iodo, pubchem14957, acmc-209qnq, 5-chloro-2-iodo pyrimidine, ksc493q5d
C1=C(C=NC(=N1)I)Cl
240.428
240.43
98%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C