Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-iodoaniline, 98%

$17.51 - $17.51

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-iodoaniline
CAS 6828-35-9
Molecular Formula C6H5ClIN
Formula Weight 253.47
MDL Number MFCD06738960
UN Number UN2811
Synonym 2-Amino-4-chloroiodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2570903

  SDS
alfa aesar™
B2570903
1g Each for $17.51

AAB2570906

  SDS
alfa aesar™
B2570906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 253.47
Assay 98%
Molecular Formula C6H5ClIN
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 2-Amino-4-chloroiodobenzene
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
MDL Number MFCD06738960
Chemical Name or Material 5-Chloro-2-iodoaniline
CAS 6828-35-9
Melting Point 40° to 42°C
UN Number UN2811
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.