missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2-hydroxypyridine, TCI America™

$30.56 - $94.72

Chemical Identifiers

CAS 4214-79-3
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.543
MDL Number MFCD00006275
InChI Key SZFUWUOHDRMCKD-UHFFFAOYSA-N
Synonym 5-chloro-2-hydroxypyridine, 5-chloropyridin-2-ol, 5-chloro-2-pyridinol, 5-chloropyridin-2 1h-one, 2-hydroxy-5-chloropyridine, 5-chloro-2-pyridol, 5-chloro-2 1h-pyridinone, 5-chloro-2-pyridone, 2 1h-pyridinone, 5-chloro, unii-r7a6f5d3ky
PubChem CID 77889
IUPAC Name 5-chloro-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C12455G View Documents TCI America
C12455G
5g Each for $30.56
 
C124525G View Documents TCI America
C124525G
25g Each for $94.72
 
Specifications

Chemical Identifiers

4214-79-3
129.543
SZFUWUOHDRMCKD-UHFFFAOYSA-N
77889
C1=CC(=O)NC=C1Cl
C5H4ClNO
MFCD00006275
5-chloro-2-hydroxypyridine, 5-chloropyridin-2-ol, 5-chloro-2-pyridinol, 5-chloropyridin-2 1h-one, 2-hydroxy-5-chloropyridine, 5-chloro-2-pyridol, 5-chloro-2 1h-pyridinone, 5-chloro-2-pyridone, 2 1h-pyridinone, 5-chloro, unii-r7a6f5d3ky
5-chloro-1H-pyridin-2-one

Specifications

White-Yellow
5g
C5H4ClNO
5-chloro-2-hydroxypyridine, 5-chloropyridin-2-ol, 5-chloro-2-pyridinol, 5-chloropyridin-2 1h-one, 2-hydroxy-5-chloropyridine, 5-chloro-2-pyridol, 5-chloro-2 1h-pyridinone, 5-chloro-2-pyridone, 2 1h-pyridinone, 5-chloro, unii-r7a6f5d3ky
C1=CC(=O)NC=C1Cl
129.543
129.54
≥98.0% (GC,T)
164°C
4214-79-3
MFCD00006275
SZFUWUOHDRMCKD-UHFFFAOYSA-N
5-chloro-1H-pyridin-2-one
77889
Crystal-Powder at 20°C
5-Chloro-2-hydroxypyridine
Safety and Handling

Safety and Handling

TSCA : No