Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-hydroxypyridine, 98+%

$36.77 - $513.81

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-hydroxypyridine
CAS 4214-79-3
Molecular Formula C5H4ClNO
Formula Weight 129.55
MDL Number MFCD00006275
Beilstein 2020
Synonym 5-Chloro-2-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1191409

  SDS
alfa aesar™
A1191409
10g Each for $36.77

AAA1191430

  SDS
alfa aesar™
A1191430
250g Each for $513.81
Description & Specifications

Specifications

Formula Weight 129.55
CAS 4214-79-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Chemical Name or Material 5-Chloro-2-hydroxypyridine
Molecular Formula C5H4ClNO
Melting Point 163° to 165°C
Quantity 10g
Health Hazard 1 Warning
MDL Number MFCD00006275
Synonym 5-Chloro-2-pyridinol
Beilstein 2020
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.