missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-hydroxypyridine, 98+%

Manufacturer:  Alfa Aesar™ A1191418

 View more versions of this product

Catalog No. AAA1191418



Specifications

Specifications

5-Chloro-2-hydroxypyridine
50g
C5H4ClNO
2020
SZFUWUOHDRMCKD-UHFFFAOYSA-N
5-chloro-1H-pyridin-2-one
77889
≥98%
163°C to 165°C
4214-79-3
MFCD00006275
5-chloro-2-hydroxypyridine, 5-chloropyridin-2-ol, 5-chloro-2-pyridinol, 5-chloropyridin-2 1h-one, 2-hydroxy-5-chloropyridine, 5-chloro-2-pyridol, 5-chloro-2 1h-pyridinone, 5-chloro-2-pyridone, 2 1h-pyridinone, 5-chloro, unii-r7a6f5d3ky
C1=CC(=O)NC=C1Cl
129.543
129.55

Chemical Identifiers

4214-79-3
129.543
SZFUWUOHDRMCKD-UHFFFAOYSA-N
77889
C1=CC(=O)NC=C1Cl
C5H4ClNO
MFCD00006275
5-chloro-2-hydroxypyridine, 5-chloropyridin-2-ol, 5-chloro-2-pyridinol, 5-chloropyridin-2 1h-one, 2-hydroxy-5-chloropyridine, 5-chloro-2-pyridol, 5-chloro-2 1h-pyridinone, 5-chloro-2-pyridone, 2 1h-pyridinone, 5-chloro, unii-r7a6f5d3ky
5-chloro-1H-pyridin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents