missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-chloro-2-hydroxynicotinic acid, 97%, Maybridge

$14.00 - $74.52

Chemical Identifiers

CAS 38076-80-1
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C6H4ClNO3
Formula Weight 173.55
MDL Number MFCD00204158
Chemical Name or Material 5-chloro-2-hydroxynicotinic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
SPB06836DA SDS Fisher Scientific Company
SPB06836DA
Amber Glass Bottle 1g Each for $14.00
SPB06836EA SDS Fisher Scientific Company
SPB06836EA
Amber Glass Bottle 10g Each for $74.52
Specifications Description & Specifications

Chemical Identifiers

38076-80-1
97%
173.55
5-chloro-2-hydroxynicotinic acid
97.0
C6H4ClNO3
MFCD00204158
97%

Specifications

173.55
97.0
MFCD00204158
Amber Glass Bottle
5-chloro-2-hydroxynicotinic acid
1g
C6H4ClNO3
97%
38076-80-1
97%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.