Please login to your online account to display your discounted pricing

5-chloro-2-hydroxynicotinic acid, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB06836EA

 View more versions of this product

Catalog No. SPB06836EA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 173.55
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 5-chloro-2-hydroxynicotinic acid
Assay Percent Range 97%
Molecular Formula C6H4ClNO3
CAS 38076-80-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Amber Glass Bottle
MDL Number MFCD00204158
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-chloro-2-hydroxynicotinic acid
CAS 38076-80-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H4ClNO3
Formula Weight 173.55
MDL Number MFCD00204158
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.