Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-hydroxynicotinic acid, 97%

$93.27 - $1,387.73

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-hydroxynicotinic acid
CAS 38076-80-1
Molecular Formula C6H4ClNO3
Formula Weight 173.56
MDL Number MFCD00204158
Synonym 5-Chloro-2-hydroxypyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2070603

  SDS
alfa aesar™
B2070603
1g Each for $93.27

AAB2070606

  SDS
alfa aesar™
B2070606
5g Each for $334.50

AAB2070614

  SDS
alfa aesar™
B2070614
25g Each for $1,387.73
Description & Specifications

Specifications

Formula Weight 173.56
Chemical Name or Material 5-Chloro-2-hydroxynicotinic acid
Assay 97%
Molecular Formula C6H4ClNO3
MDL Number MFCD00204158
CAS 38076-80-1
Synonym 5-Chloro-2-hydroxypyridine-3-carboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 258° to 260°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.