Learn More
Alfa Aesar™ 5-Chloro-2-hydroxybenzeneboronic acid, 97%
Manufacturer: Alfa Aesar™ H5307503
Specifications
5-Chloro-2-hydroxybenzeneboronic acid | |
1g | |
C6H6BClO3 | |
5-chloro-2-hydroxybenzeneboronic acid, 2-borono-4-chlorophenol, 5-chloro-2-hydroxyphenyl boronic acid, 5-chloro-2-hydroxyphenyl-boronic acid, boronic acid, 5-chloro-2-hydroxyphenyl, pubchem19653, acmc-209r1f, 2-dihydroxyboryl-4-chlorophenol, 5-chloro-2-hydroxyphenyl boronic acid, boronic acid,b-5-chloro-2-hydroxyphenyl | |
B(C1=C(C=CC(=C1)Cl)O)(O)O | |
172.371 | |
172.37 |
166°C to 170°C | |
89488-25-5 | |
MFCD06659863 | |
GGZASRFCRAZNPO-UHFFFAOYSA-N | |
(5-chloro-2-hydroxyphenyl)boronic acid | |
23005362 | |
97% |
Chemical Identifiers
89488-25-5 | |
172.371 | |
GGZASRFCRAZNPO-UHFFFAOYSA-N | |
23005362 | |
B(C1=C(C=CC(=C1)Cl)O)(O)O |
C6H6BClO3 | |
MFCD06659863 | |
5-chloro-2-hydroxybenzeneboronic acid, 2-borono-4-chlorophenol, 5-chloro-2-hydroxyphenyl boronic acid, 5-chloro-2-hydroxyphenyl-boronic acid, boronic acid, 5-chloro-2-hydroxyphenyl, pubchem19653, acmc-209r1f, 2-dihydroxyboryl-4-chlorophenol, 5-chloro-2-hydroxyphenyl boronic acid, boronic acid,b-5-chloro-2-hydroxyphenyl | |
(5-chloro-2-hydroxyphenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning