Please login to your online account to display your discounted pricing

Alfa Aesar™ 5'-Chloro-2'-hydroxyacetophenone, 99%

$18.62 - $34.64

Chemical Identifiers

Chemical Name or Material 5'-Chloro-2'-hydroxyacetophenone
CAS 1450-74-4
Molecular Formula C8H7ClO2
Formula Weight 170.6
MDL Number MFCD00067788
Beilstein 1365589
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1544803

 
alfa aesar™
A1544803
1g Each for $18.62

AAA1544806

 
alfa aesar™
A1544806
5g Each for $34.64
Description & Specifications

Specifications

Formula Weight 170.6
Chemical Name or Material 5'-Chloro-2'-hydroxyacetophenone
Assay 99%
Molecular Formula C8H7ClO2
MDL Number MFCD00067788
CAS 1450-74-4
Beilstein 1365589
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 107° to 109°C (12mmHg)
Melting Point 52° to 53°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.