Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-hydroxy-3-nitropyridine, 97%

$61.47 - $785.03

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-hydroxy-3-nitropyridine
CAS 21427-61-2
Molecular Formula C5H3ClN2O3
Formula Weight 174.54
MDL Number MFCD00114884
Beilstein 383852
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1986003

  SDS
alfa aesar™
L1986003
1g Each for $61.47

AAL1986014

  SDS
alfa aesar™
L1986014
25g Each for $785.03

AAL1986006

  SDS
alfa aesar™
L1986006
5g Each for $201.91
Description & Specifications

Specifications

Formula Weight 174.54
Chemical Name or Material 5-Chloro-2-hydroxy-3-nitropyridine
Assay 97%
Molecular Formula C5H3ClN2O3
MDL Number MFCD00114884
CAS 21427-61-2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 383852
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 234° to 236°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.