Please login to your online account to display your discounted pricing

Alfa Aesar™ 5'-Chloro-2'-hydroxy-3'-nitroacetophenone, 99%

$51.38 - $168.76

Chemical Identifiers

Chemical Name or Material 5'-Chloro-2'-hydroxy-3'-nitroacetophenone
CAS 84942-40-5
Molecular Formula C8H6ClNO4
Formula Weight 215.6
MDL Number MFCD00191921
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2741706

  SDS
alfa aesar™
H2741706
5g Each for $51.38

AAH2741714

  SDS
alfa aesar™
H2741714
25g Each for $168.76
Description & Specifications

Specifications

Formula Weight 215.6
Chemical Name or Material 5'-Chloro-2'-hydroxy-3'-nitroacetophenone
Assay 99%
Molecular Formula C8H6ClNO4
MDL Number MFCD00191921
CAS 84942-40-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 132° to 135°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.