Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-hydroxy-3-methylbenzaldehyde, 97%

$455.04 - $455.04

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-hydroxy-3-methylbenzaldehyde
CAS 23602-63-3
Molecular Formula C8H7ClO2
Formula Weight 170.6
MDL Number MFCD00275687
Beilstein 2249708
Synonym 5-Chloro-3-methylsalicylaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1979306

  SDS
alfa aesar™
L1979306
5g Each for $455.04
Description & Specifications

Specifications

Formula Weight 170.6
Molecular Formula C8H7ClO2
Synonym 5-Chloro-3-methylsalicylaldehyde
Assay 97%
MDL Number MFCD00275687
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 23602-63-3
Melting Point 59°C
Health Hazard 1 Warning
Chemical Name or Material 5-Chloro-2-hydroxy-3-methylbenzaldehyde
Quantity 5g
Beilstein 2249708
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.