Please login to your online account to display your discounted pricing

5-Chloro-2-hydroxy-3-methylbenzaldehyde, 95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB03914CB

 View more versions of this product

Catalog No. BTB03914CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 170.59
Quantity 250mg
Assay 95%
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C8H7ClO2
Packaging Amber Glass Bottle
MDL Number MFCD00275687
CAS 23602-63-3
CAS Min % 94.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 5-Chloro-2-hydroxy-3-methylbenzaldehyde

Specifications

Chemical Name or Material 5-Chloro-2-hydroxy-3-methylbenzaldehyde
CAS 23602-63-3
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H7ClO2
Formula Weight 170.59
MDL Number MFCD00275687
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.