Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-hydrazino-3-iodopyridine, 95%

$153.93 - $347.88

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-hydrazino-3-iodopyridine
CAS 942206-12-4
Molecular Formula C5H5ClIN3
Formula Weight 269.47
MDL Number MFCD09037446
Synonym (5-Chloro-3-iodo-2-pyridyl)hydrazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2711703

 
alfa aesar™
H2711703
1g Each for $347.88

AAH27117MD

 
alfa aesar™
H27117MD
250mg Each for $153.93
Description & Specifications

Specifications

Formula Weight 269.47
Chemical Name or Material 5-Chloro-2-hydrazino-3-iodopyridine
Assay 95%
Molecular Formula C5H5ClIN3
MDL Number MFCD09037446
CAS 942206-12-4
Synonym (5-Chloro-3-iodo-2-pyridyl)hydrazine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 136° to 138°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.