Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-furaldehyde, 98%, stab. with 2% ethanol

$60.03 - $60.03

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-furaldehyde
CAS 21508-19-0
Name Note stabilized with 2% ethanol
Molecular Formula C5H3ClO2
Formula Weight 130.53
MDL Number MFCD02752599
Beilstein 108401
Synonym 5-Chlorofuran-2-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1997103

 
alfa aesar™
L1997103
1g Each for $60.03
Description & Specifications

Specifications

Formula Weight 130.53
MDL Number MFCD02752599
Synonym 5-Chlorofuran-2-carboxaldehyde
Name Note stabilized with 2% ethanol
Molecular Formula C5H3ClO2
Beilstein 108401
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Boiling Point 70°C (10mmHg)
Melting Point 33° to 36°C
Quantity 1g
Chemical Name or Material 5-Chloro-2-furaldehyde
Assay 98%
CAS 21508-19-0

Applications
(+)-1-Deoxynojirimycin hydrochloride interferes with normal processing of N-linked glycoproteins, producing reduced amounts of complex type oligosaccharides.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C