Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-fluorotoluene, 98%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-fluorotoluene
CAS 452-66-4
Molecular Formula C7H6ClF
Formula Weight 144.58
MDL Number MFCD00018504
UN Number UN1993
Beilstein 1931799
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0761506

  SDS
alfa aesar™
L0761506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0761514

  SDS
alfa aesar™
L0761514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.58
Molecular Formula C7H6ClF
CAS 452-66-4
MDL Number MFCD00018504
Chemical Name or Material 5-Chloro-2-fluorotoluene
Health Hazard 3 GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Warning
Boiling Point 157° to 159°C
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Assay 98%
UN Number UN1993
Flash Point 48°C (118°F)
Odor Aromatic
Beilstein 1931799
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.