Please login to your online account to display your discounted pricing

5-chloro-2-fluorophenyl isothiocyanate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-fluorophenyl isothiocyanate
CAS 247170-25-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H3ClFNS
Formula Weight 187.62
MDL Number MFCD01313224
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01971EA

  SDS
fisher scientific company
SB01971EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.62
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Molecular Formula C7H3ClFNS
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
MDL Number MFCD01313224
Chemical Name or Material 5-Chloro-2-fluorophenyl isothiocyanate
CAS 247170-25-8
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.