Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-fluorobenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-fluorobenzyl bromide
CAS 71916-91-1
Molecular Formula C7H5BrClF
Formula Weight 223.48
MDL Number MFCD01631437
UN Number UN3265
Beilstein 637765
Synonym alpha-Bromo-3-chloro-6-fluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1914703

  SDS
alfa aesar™
L1914703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL19147MD

  SDS
alfa aesar™
L19147MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 223.48
Molecular Formula C7H5BrClF
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
MDL Number MFCD01631437
Health Hazard 1 Danger
Chemical Name or Material 5-Chloro-2-fluorobenzyl bromide
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Quantity 1g
UN Number UN3265
CAS 71916-91-1
Assay 97%
Beilstein 637765
Boiling Point 44° to 45°C (0.5mmHg)
Synonym alpha-Bromo-3-chloro-6-fluorotoluene
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.