missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Chloro-2-fluorobenzoyl chloride, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC442500050
Specifications
5-Chloro-2-fluorobenzoyl chloride | |
100°C | |
103.0°C to 104.0°C (15.0 Torr) | |
394-29-6 | |
C7H3Cl2FO | |
VWGAATKTEAKOCO-UHFFFAOYSA-N | |
5-chloro-2-fluorobenzoyl chloride | |
2773604 | |
97% |
1.4590g/mL | |
1.459 | |
5mL | |
96% min. (GC) | |
2-fluoro-5-chlorobenzoyl chloride, 5-chloranyl-2-fluoranyl-benzoyl chloride, 5-chloro-2-fluoro-benzoyl chloride, 5-chloro-2-fluorobenzoyl chloride, 5-chloro-2-fluorobenzoylchloride, acmc-1aho4, benzoyl chloride, 5-chloro-2-fluoro, benzoyl chloride,5-chloro-2-fluoro, buttpark 45\01-10 | |
C1=CC(=C(C=C1Cl)C(=O)Cl)F | |
193.01 | |
193.01 |
Chemical Identifiers
394-29-6 | |
193.01 | |
2-fluoro-5-chlorobenzoyl chloride, 5-chloranyl-2-fluoranyl-benzoyl chloride, 5-chloro-2-fluoro-benzoyl chloride, 5-chloro-2-fluorobenzoyl chloride, 5-chloro-2-fluorobenzoylchloride, acmc-1aho4, benzoyl chloride, 5-chloro-2-fluoro, benzoyl chloride,5-chloro-2-fluoro, buttpark 45\01-10 | |
5-chloro-2-fluorobenzoyl chloride |
C7H3Cl2FO | |
VWGAATKTEAKOCO-UHFFFAOYSA-N | |
2773604 | |
C1=CC(=C(C=C1Cl)C(=O)Cl)F |
Safety and Handling
GHS Signal Word: Danger