Please login to your online account to display your discounted pricing

5-Chloro-2-fluorobenzoyl chloride, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-fluorobenzoyl chloride
Assay Percent Range 97%
Molecular Formula C7H3Cl2FO
Formula Weight 193.01
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC442500010

 
thermo fisher scientific
AC442500010
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442500050

 
thermo fisher scientific
AC442500050
5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.01
Quantity 1mL
Chemical Name or Material 5-Chloro-2-fluorobenzoyl chloride
CAS 394-29-6
Molecular Formula C7H3Cl2FO
Assay Percent Range 97%