Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-fluorobenzotrifluoride, 98+%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-fluorobenzotrifluoride
CAS 89634-74-2
Molecular Formula C7H3ClF4
Formula Weight 198.55
MDL Number MFCD00060655
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2620809

  SDS
alfa aesar™
H2620809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198.55
UN Number UN1993
Quantity 10g
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00060655
Boiling Point 145°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Chemical Name or Material 5-Chloro-2-fluorobenzotrifluoride
Flash Point 47°C (117°F)
Assay ≥98%
Molecular Formula C7H3ClF4
CAS 89634-74-2
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.