Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-fluorobenzoic acid, 97%

$206.43 - $206.43

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-fluorobenzoic acid
CAS 394-30-9
Molecular Formula C7H4ClFO2
Formula Weight 174.56
MDL Number MFCD00665762
Beilstein 2614286
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2377606

  SDS
alfa aesar™
B2377606
5g Each for $206.43
Description & Specifications

Specifications

Formula Weight 174.56
Chemical Name or Material 5-Chloro-2-fluorobenzoic acid
Assay 97%
Molecular Formula C7H4ClFO2
MDL Number MFCD00665762
CAS 394-30-9
Beilstein 2614286
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 148° to 150°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.