missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-fluorobenzenesulfonamide, 97%

$90.53 - $299.18

Chemical Identifiers

CAS 351003-57-1
Molecular Formula C6H5ClFNO2S
Molecular Weight (g/mol) 209.619
MDL Number MFCD03094289
InChI Key DBJNLJWQSQOBJM-UHFFFAOYSA-N
Synonym 5-chloranyl-2-fluoranyl-benzenesulfonamide
PubChem CID 1268284
IUPAC Name 5-chloro-2-fluorobenzenesulfonamide
SMILES C1=CC(=C(C=C1Cl)S(=O)(=O)N)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3214403 View Documents Alfa Aesar™
H3214403
1g Each for $90.53
 
AAH3214406 View Documents Alfa Aesar™
H3214406
5g Each for $299.18
 
Specifications

Chemical Identifiers

351003-57-1
209.619
DBJNLJWQSQOBJM-UHFFFAOYSA-N
1268284
C1=CC(=C(C=C1Cl)S(=O)(=O)N)F
C6H5ClFNO2S
MFCD03094289
5-chloranyl-2-fluoranyl-benzenesulfonamide
5-chloro-2-fluorobenzenesulfonamide

Specifications

134°C to 136°C
351003-57-1
MFCD03094289
DBJNLJWQSQOBJM-UHFFFAOYSA-N
5-chloro-2-fluorobenzenesulfonamide
1268284
97%
1g
C6H5ClFNO2S
5-chloranyl-2-fluoranyl-benzenesulfonamide
C1=CC(=C(C=C1Cl)S(=O)(=O)N)F
209.619
209.63
5-Chloro-2-fluorobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning