missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5310803

 View more versions of this product

Catalog No. AAH5310803



Specifications

Specifications

5-Chloro-2-fluorobenzeneboronic acid
1g
C6H5BClFO2
5-chloro-2-fluorophenyl boronic acid, 5-chloro-2-fluorobenzeneboronic acid, 5-chloro-2-fluorophenylboronicacid, 5-chloro-2-fluoro-phenyl boronic acid, boronic acid, 5-chloro-2-fluorophenyl, pubchem11819, acmc-1ae0a, 5-chloro-2-fluorophenyl boronic acid, boronic acid,b-5-chloro-2-fluorophenyl, boronic acid, b-5-chloro-2-fluorophenyl
B(C1=C(C=CC(=C1)Cl)F)(O)O
174.362
174.37
182°C to 188°C
352535-83-2
MFCD05664225
GGTUVWGMCFXUAS-UHFFFAOYSA-N
(5-chloro-2-fluorophenyl)boronic acid
3718925
97%

Chemical Identifiers

352535-83-2
174.362
GGTUVWGMCFXUAS-UHFFFAOYSA-N
3718925
B(C1=C(C=CC(=C1)Cl)F)(O)O
C6H5BClFO2
MFCD05664225
5-chloro-2-fluorophenyl boronic acid, 5-chloro-2-fluorobenzeneboronic acid, 5-chloro-2-fluorophenylboronicacid, 5-chloro-2-fluoro-phenyl boronic acid, boronic acid, 5-chloro-2-fluorophenyl, pubchem11819, acmc-1ae0a, 5-chloro-2-fluorophenyl boronic acid, boronic acid,b-5-chloro-2-fluorophenyl, boronic acid, b-5-chloro-2-fluorophenyl
(5-chloro-2-fluorophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents