Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-fluorobenzamide, 97+%

$78.81 - $280.15

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-fluorobenzamide
CAS 261762-57-6
Molecular Formula C7H5ClFNO
Formula Weight 173.58
MDL Number MFCD01631435
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2462304

  SDS
alfa aesar™
B2462304
2g Each for $78.81

AAB2462309

  SDS
alfa aesar™
B2462309
10g Each for $280.15
Description & Specifications

Specifications

Formula Weight 173.58
Chemical Name or Material 5-Chloro-2-fluorobenzamide
Assay ≥97%
Molecular Formula C7H5ClFNO
MDL Number MFCD01631435
CAS 261762-57-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 134° to 138°C
Quantity 2g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.