missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-ethoxybenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5331206

 View more versions of this product

Catalog No. AAH5331206



Specifications

Specifications

5-Chloro-2-ethoxybenzeneboronic acid
5g
C8H10BClO3
5-chloro-2-ethoxybenzeneboronic acid, 2-ethoxy-5-chlorophenylboronic acid, 5-chloro-2-ethoxyphenyl boronic acid, 5-chloro-2-ethoxypheylboronic acid, boronic acid, 5-chloro-2-ethoxyphenyl, 5-chloro-2-ethoxy-phenyl boronic acid, acmc-209if6, 5-chloro-2-ethoxypheylboronicacid, 2-ethoxy-5-chloro-phenylboronic acid, 5-chloro-2-ethoxy-phenylboronic acid
B(C1=C(C=CC(=C1)Cl)OCC)(O)O
200.425
200.43
110°C to 114°C
352534-86-2
MFCD03427055
OSMBQNBPCSSVMT-UHFFFAOYSA-N
(5-chloro-2-ethoxyphenyl)boronic acid
3700836
97%

Chemical Identifiers

352534-86-2
200.425
OSMBQNBPCSSVMT-UHFFFAOYSA-N
3700836
B(C1=C(C=CC(=C1)Cl)OCC)(O)O
C8H10BClO3
MFCD03427055
5-chloro-2-ethoxybenzeneboronic acid, 2-ethoxy-5-chlorophenylboronic acid, 5-chloro-2-ethoxyphenyl boronic acid, 5-chloro-2-ethoxypheylboronic acid, boronic acid, 5-chloro-2-ethoxyphenyl, 5-chloro-2-ethoxy-phenyl boronic acid, acmc-209if6, 5-chloro-2-ethoxypheylboronicacid, 2-ethoxy-5-chloro-phenylboronic acid, 5-chloro-2-ethoxy-phenylboronic acid
(5-chloro-2-ethoxyphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents