missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Chloro-2-cyanopyridine 98.0+%, TCI America™
Supplier: TCI America C302325G
Specifications
5-Chloro-2-cyanopyridine | |
109°C | |
110°C | |
MFCD03788835 | |
3439 | |
WTHODOKFSYPTKA-UHFFFAOYSA-N | |
5-chloropyridine-2-carbonitrile | |
3833942 | |
≥98.0% (GC) |
89809-64-3 | |
Yellow | |
C6H3ClN2 | |
25 g | |
5-chloro-2-cyanopyridine, 5-chloropicolinonitrile, 3-chloro-6-cyanopyridine, 2-cyano-5-chloro pyridine, 2-pyridinecarbonitrile, 5-chloro, 2-cyano-5-chloropyridine, 6-cyano-3-chloropyridine, 5-chloro-2-pyridinecarbonitrile, pubchem2107, 5-chloropicolinenitrile | |
ClC1=CN=C(C=C1)C#N | |
138.55 | |
138.55 | |
Crystalline Powder |
Chemical Identifiers
89809-64-3 | |
138.55 | |
WTHODOKFSYPTKA-UHFFFAOYSA-N | |
3833942 | |
ClC1=CN=C(C=C1)C#N |
C6H3ClN2 | |
MFCD03788835 | |
5-chloro-2-cyanopyridine, 5-chloropicolinonitrile, 3-chloro-6-cyanopyridine, 2-cyano-5-chloro pyridine, 2-pyridinecarbonitrile, 5-chloro, 2-cyano-5-chloropyridine, 6-cyano-3-chloropyridine, 5-chloro-2-pyridinecarbonitrile, pubchem2107, 5-chloropicolinenitrile | |
5-chloropyridine-2-carbonitrile |
Safety and Handling
TSCA : No