missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2-cyanopyridine, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6484606

 View more versions of this product

Catalog No. AAH6484606



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Chloro-2-cyanopyridine
106°C to 108°C
MFCD03788835
UN3439
5-chloro-2-cyanopyridine, 5-chloropicolinonitrile, 3-chloro-6-cyanopyridine, 2-cyano-5-chloro pyridine, 2-pyridinecarbonitrile, 5-chloro, 2-cyano-5-chloropyridine, 6-cyano-3-chloropyridine, 5-chloro-2-pyridinecarbonitrile, pubchem2107, 5-chloropicolinenitrile
ClC1=CN=C(C=C1)C#N
138.55
138.55
89809-64-3
C6H3ClN2
5 g
113764
WTHODOKFSYPTKA-UHFFFAOYSA-N
5-chloropyridine-2-carbonitrile
3833942
96%

Chemical Identifiers

89809-64-3
138.55
WTHODOKFSYPTKA-UHFFFAOYSA-N
3833942
ClC1=CN=C(C=C1)C#N
C6H3ClN2
MFCD03788835
5-chloro-2-cyanopyridine, 5-chloropicolinonitrile, 3-chloro-6-cyanopyridine, 2-cyano-5-chloro pyridine, 2-pyridinecarbonitrile, 5-chloro, 2-cyano-5-chloropyridine, 6-cyano-3-chloropyridine, 5-chloro-2-pyridinecarbonitrile, pubchem2107, 5-chloropicolinenitrile
5-chloropyridine-2-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only