Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-cyanobenzeneboronic acid, 98%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-cyanobenzeneboronic acid
CAS 1072946-52-1
Formula Weight 225.84
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6184203

 
alfa aesar™
H6184203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61842MD

 
alfa aesar™
H61842MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.84
Chemical Name or Material 5-Chloro-2-cyanobenzeneboronic acid
Physical Form Solid
Assay 98%
CAS 1072946-52-1
Quantity 1g