Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-adamantanone, 90+%

$439.97 - $439.97

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-adamantanone
CAS 20098-17-3
Molecular Formula C10H13ClO
Formula Weight 184.67
MDL Number MFCD00798599
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2102806

  SDS
alfa aesar™
B2102806
5g Each for $439.97
Description & Specifications

Specifications

Formula Weight 184.67
CAS 20098-17-3
Quantity 5g
MDL Number MFCD00798599
Chemical Name or Material 5-Chloro-2-adamantanone
Assay ≥90%
Molecular Formula C10H13ClO