missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2,4-dinitrotoluene, 97%

Manufacturer:  Alfa Aesar™ L1180814

 View more versions of this product

Catalog No. AAL1180814



Specifications

Specifications

5-Chloro-2,4-dinitrotoluene
25g
C7H5ClN2O4
UN2811
5-chloro-2,4-dinitrotoluene, toluene, 5-chloro-2,4-dinitro, 1-chloro-5-methyl-2,4-dinitro-benzene, 2-chloro-4-methyl-1,5-dinitrobenzene, acmc-20am4s, 5-chlor-2.4-dinitrotoluol, 5-chloro-2,4-dinitrotoluene, benzene,1-chloro-5-methyl-2,4-dinitro, 1-chloranyl-5-methyl-2,4-dinitro-benzene
CC1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])Cl
216.577
216.58
89°C to 91°C
51676-74-5
MFCD00041235
2506591
KPDPGZNHKMJEFZ-UHFFFAOYSA-N
1-chloro-5-methyl-2,4-dinitrobenzene
253802
97%

Chemical Identifiers

51676-74-5
216.577
KPDPGZNHKMJEFZ-UHFFFAOYSA-N
253802
CC1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])Cl
C7H5ClN2O4
MFCD00041235
5-chloro-2,4-dinitrotoluene, toluene, 5-chloro-2,4-dinitro, 1-chloro-5-methyl-2,4-dinitro-benzene, 2-chloro-4-methyl-1,5-dinitrobenzene, acmc-20am4s, 5-chlor-2.4-dinitrotoluol, 5-chloro-2,4-dinitrotoluene, benzene,1-chloro-5-methyl-2,4-dinitro, 1-chloranyl-5-methyl-2,4-dinitro-benzene
1-chloro-5-methyl-2,4-dinitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents