missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2,4-dinitrobenzoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2547906

 View more versions of this product

Catalog No. AAB2547906



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Chloro-2,4-dinitrobenzoic acid
180°C to 182°C
MFCD00091829
5-chloro-2,4-dinitrobenzoic acid, benzoic acid, 5-chloro-2,4-dinitro, acmc-20a30o, 2,4-dinitro-5-chlorobenzoic acid, 3-chloro-4,6-dinitrobenzoic acid
C1=C(C(=CC(=C1Cl)[N+](=O)[O-])[N+](=O)[O-])C(=O)O
3727860
98%
136833-36-8
C7H3ClN2O6
5 g
VVQOEJCXWMGEJG-UHFFFAOYSA-N
246.559
246.56

Chemical Identifiers

136833-36-8
246.559
VVQOEJCXWMGEJG-UHFFFAOYSA-N
3727860
C7H3ClN2O6
MFCD00091829
5-chloro-2,4-dinitrobenzoic acid, benzoic acid, 5-chloro-2,4-dinitro, acmc-20a30o, 2,4-dinitro-5-chlorobenzoic acid, 3-chloro-4,6-dinitrobenzoic acid
C1=C(C(=CC(=C1Cl)[N+](=O)[O-])[N+](=O)[O-])C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only