Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2-[4-(dimethylamino)phenyl]benzimidazole, 95%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2-[4-(dimethylamino)phenyl]benzimidazole
CAS 1030574-44-7
Formula Weight 271.75
MDL Number MFCD15173053
Beilstein 22393779
Synonym 4-(5-Chloro-2-benzimidazolyl)-N,N-dimethylaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6499203

  SDS
alfa aesar™
H6499203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6499206

  SDS
alfa aesar™
H6499206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64992MD

  SDS
alfa aesar™
H64992MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 271.75
Chemical Name or Material 5-Chloro-2-[4-(dimethylamino)phenyl]benzimidazole
Assay 95%
CAS 1030574-44-7
Synonym 4-(5-Chloro-2-benzimidazolyl)-N,N-dimethylaniline
Physical Form Solid
Beilstein 22393779
Quantity 1g
MDL Number MFCD15173053