missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2,4-dimethoxyaniline, TCI America™

$50.42 - $380.42

Chemical Identifiers

CAS 97-50-7
Molecular Formula C8H10ClNO2
Molecular Weight (g/mol) 187.623
MDL Number MFCD00025752
InChI Key OLCMNCWEUMBNIS-UHFFFAOYSA-N
Synonym itr amine, benzenamine, 5-chloro-2,4-dimethoxy, 2,4-dimethoxy-5-chloroaniline, 5-chloro-2,4-dimethoxyaminobenzene, unii-lz646979la, aniline,2,4-dimethoxy-5-chloro, 5-chloro-2,4-dimethoxyphenylamine, acmc-209s8h, cambridge id 5252837, ksc496a0p
PubChem CID 66807
IUPAC Name 5-chloro-2,4-dimethoxyaniline
SMILES COC1=CC(=C(C=C1N)Cl)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C122625G View Documents TCI America
C122625G
25g Each for $50.42
 
C1226500G View Documents TCI America
C1226500G
500g Each for $380.42
 
Specifications

Chemical Identifiers

97-50-7
187.623
OLCMNCWEUMBNIS-UHFFFAOYSA-N
66807
COC1=CC(=C(C=C1N)Cl)OC
C8H10ClNO2
MFCD00025752
itr amine, benzenamine, 5-chloro-2,4-dimethoxy, 2,4-dimethoxy-5-chloroaniline, 5-chloro-2,4-dimethoxyaminobenzene, unii-lz646979la, aniline,2,4-dimethoxy-5-chloro, 5-chloro-2,4-dimethoxyphenylamine, acmc-209s8h, cambridge id 5252837, ksc496a0p
5-chloro-2,4-dimethoxyaniline

Specifications

Purple
25g
C8H10ClNO2
itr amine, benzenamine, 5-chloro-2,4-dimethoxy, 2,4-dimethoxy-5-chloroaniline, 5-chloro-2,4-dimethoxyaminobenzene, unii-lz646979la, aniline,2,4-dimethoxy-5-chloro, 5-chloro-2,4-dimethoxyphenylamine, acmc-209s8h, cambridge id 5252837, ksc496a0p
COC1=CC(=C(C=C1N)Cl)OC
187.623
187.62
≥97.0% (GC,T)
92°C
97-50-7
MFCD00025752
OLCMNCWEUMBNIS-UHFFFAOYSA-N
5-chloro-2,4-dimethoxyaniline
66807
Crystal-Powder at 20°C
5-Chloro-2,4-dimethoxyaniline
Safety and Handling

Safety and Handling

TSCA : Yes