Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2,4-dimethoxyaniline, 98%

Chemical Identifiers

Chemical Name or Material 5-Chloro-2,4-dimethoxyaniline
CAS 97-50-7
Formula Weight 187.63
MDL Number MFCD00025752
UN Number UN3077
Beilstein 2211013
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5921403

  SDS
alfa aesar™
H5921403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.63
Melting Point 87° to 90°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Physical Form Solid
TSCA TSCA
UN Number UN3077
Beilstein 2211013
Assay 98%
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
CAS 97-50-7
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00025752
Color Paple purple
Quantity 1g
Chemical Name or Material 5-Chloro-2,4-dimethoxyaniline
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.