Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-2(3H)-benzoxazolone, 99%

$76.39 - $780.50

Chemical Identifiers

Chemical Name or Material 5-Chloro-2(3H)-benzoxazolone
CAS 95-25-0
Molecular Formula C7H4ClNO2
Formula Weight 169.57
MDL Number MFCD00005717
Merck Index 14,2194
Synonym 5-Chloro-2-benzoxazolinone; Chlorzoxazone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB245070B

  SDS
alfa aesar™
B245070B
1000g Each for $780.50

AAB2450718

  SDS
alfa aesar™
B2450718
50g Each for $76.39

AAB2450730

  SDS
alfa aesar™
B2450730
250g Each for $262.18
Description & Specifications

Specifications

Formula Weight 169.57
Assay 99%
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H4ClNO2
Health Hazard 1 Danger
Quantity 1000g
Synonym 5-Chloro-2-benzoxazolinone; Chlorzoxazone
TSCA TSCA
MDL Number MFCD00005717
CAS 95-25-0
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Merck Index 14,2194
Melting Point 189° to 192°C
Chemical Name or Material 5-Chloro-2(3H)-benzoxazolone
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.