missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2,3-diphenylpyrazine 98.0+%, TCI America™
$147.25 - $736.30
Chemical Identifiers
| CAS | 41270-66-0 |
|---|---|
| Molecular Formula | C16H11ClN2 |
| Molecular Weight (g/mol) | 266.728 |
| MDL Number | MFCD00234892 |
| InChI Key | VUGNCPVAXWZTOL-UHFFFAOYSA-N |
| PubChem CID | 3654493 |
| IUPAC Name | 5-chloro-2,3-diphenylpyrazine |
| SMILES | C1=CC=C(C=C1)C2=NC=C(N=C2C3=CC=CC=C3)Cl |
Chemical Identifiers
| 41270-66-0 | |
| 266.728 | |
| VUGNCPVAXWZTOL-UHFFFAOYSA-N | |
| 5-chloro-2,3-diphenylpyrazine |
| C16H11ClN2 | |
| MFCD00234892 | |
| 3654493 | |
| C1=CC=C(C=C1)C2=NC=C(N=C2C3=CC=CC=C3)Cl |
Specifications
| 41270-66-0 | |
| White-Yellow | |
| C16H11ClN2 | |
| 1 g | |
| C1=CC=C(C=C1)C2=NC=C(N=C2C3=CC=CC=C3)Cl | |
| 266.728 | |
| 266.73 | |
| Crystalline Powder |
| 127°C | |
| 145°C | |
| MFCD00234892 | |
| VUGNCPVAXWZTOL-UHFFFAOYSA-N | |
| 5-chloro-2,3-diphenylpyrazine | |
| 3654493 | |
| ≥98.0% (GC) | |
| 5-Chloro-2,3-diphenylpyrazine |
Safety and Handling
TSCA : No