Please login to your online account to display your discounted pricing

5-chloro-2,3-dihydroxypyridine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Chloro-2, 3-dihydroxypyridine
Name Note 98%
CAS 53233-89-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5 H4 Cl N O2
Formula Weight 145.55
MDL Number MFCD00023474
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC367000050

  SDS
thermo fisher scientific
AC367000050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC367000250

  SDS
thermo fisher scientific
AC367000250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Color Grayish-beige
Physical Form Crystalline Powder
Formula Weight 145.55
Quantity 5g
CAS Max % 100.0
Chemical Name or Material 5-Chloro-2, 3-dihydroxypyridine
Packaging Glass bottle
Molecular Formula C5 H4 Cl N O2
CAS 53233-89-9
Name Note 98%
Assay Percent Range 98%
MDL Number MFCD00023474
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.