Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-1-methylimidazole, 98%

$66.91 - $650.92

Chemical Identifiers

Chemical Name or Material 5-Chloro-1-methylimidazole
CAS 872-49-1
Molecular Formula C4H5ClN2
Formula Weight 116.55
MDL Number MFCD00014505
Beilstein 110509
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1189903

  SDS
alfa aesar™
L1189903
1g Each for $168.76

AAL1189906

  SDS
alfa aesar™
L1189906
5g Each for $650.92

AAL11899MD

  SDS
alfa aesar™
L11899MD
250mg Each for $66.91
Description & Specifications

Specifications

Formula Weight 116.55
Chemical Name or Material 5-Chloro-1-methylimidazole
Assay 98%
Molecular Formula C4H5ClN2
CAS 872-49-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
MDL Number MFCD00014505
Beilstein 110509
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 96°C (204°F)
Boiling Point 89° to 90°C (14mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C