missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-1-methylimidazole, 98%

Manufacturer:  Alfa Aesar™ L1189906

 View more versions of this product

Catalog No. AAL1189906



Specifications

Specifications

5-Chloro-1-methylimidazole
89°C to 90°C (14mmHg)
872-49-1
MFCD00014505
5-chloro-1-methyl-1h-imidazole, 1-methyl-5-chloroimidazole, 1h-imidazole, 5-chloro-1-methyl, 5-chloro-1-methylimidazole, zlchem 661, pubchem7612, acmc-209qj7, 5-chloro-1-methyl imidazole, 5-chloro-1-methyl-imidazole, ksc494e7d
CN1C=NC=C1Cl
116.548
116.55
96°C (204°F)
5g
C4H5ClN2
110509
NYDGOZPYEABERA-UHFFFAOYSA-N
5-chloro-1-methylimidazole
70105
98%

Chemical Identifiers

872-49-1
116.548
NYDGOZPYEABERA-UHFFFAOYSA-N
70105
CN1C=NC=C1Cl
C4H5ClN2
MFCD00014505
5-chloro-1-methyl-1h-imidazole, 1-methyl-5-chloroimidazole, 1h-imidazole, 5-chloro-1-methyl, 5-chloro-1-methylimidazole, zlchem 661, pubchem7612, acmc-209qj7, 5-chloro-1-methyl imidazole, 5-chloro-1-methyl-imidazole, ksc494e7d
5-chloro-1-methylimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents