Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-1-indanone, 99%

$71.58 - $237.05

Chemical Identifiers

Chemical Name or Material 5-Chloro-1-indanone
CAS 42348-86-7
Molecular Formula C9H7ClO
Formula Weight 166.61
MDL Number MFCD00041456
Beilstein 1448000
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1830803

 
alfa aesar™
A1830803
1g Each for $71.58

AAA1830806

 
alfa aesar™
A1830806
5g Each for $237.05
Description & Specifications

Specifications

Formula Weight 166.61
Chemical Name or Material 5-Chloro-1-indanone
TSCA TSCA
Assay 99%
Molecular Formula C9H7ClO
MDL Number MFCD00041456
CAS 42348-86-7
Beilstein 1448000
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 124° to 125°C (3mmHg)
Melting Point 92° to 96°C
Quantity 1g

Applications
5-Chloro-1-indanone may be used as starting reagent for the preparation of 5-chloro-2-methoxycarbonyl-1-indanone. It may also be used for the preparation of important biomedical compounds such as anticonvulsants, anticholinergics and diarylsulfonylureas, having potential activity against solid tumors. It participates in the Irie′s synthesis of substituted pyridines.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.