missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-1-indanone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1830803

 View more versions of this product

Catalog No. AAA1830803



Description

Description

5-Chloro-1-indanone may be used as starting reagent for the preparation of 5-chloro-2-methoxycarbonyl-1-indanone. It may also be used for the preparation of important biomedical compounds such as anticonvulsants, anticholinergics and diarylsulfonylureas, having potential activity against solid tumors. It participates in the Irie?s synthesis of substituted pyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-1-indanone may be used as starting reagent for the preparation of 5-chloro-2-methoxycarbonyl-1-indanone. It may also be used for the preparation of important biomedical compounds such as anticonvulsants, anticholinergics and diarylsulfonylureas, having potential activity against solid tumors. It participates in the Irie′s synthesis of substituted pyridines.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

5-Chloro-1-indanone
92°C to 96°C
C9H7ClO
1 g
5-chloro-1-indanone, 5-chloro-2,3-dihydro-1h-inden-1-one, 5-chloroindan-1-one, 5-chloro-indan-1-one, 1h-inden-1-one, 5-chloro-2,3-dihydro, 5-chloroindanone, 5-chloroindone, pubchem2250, 1-indanone, 5-chloro, 5-chloro indanone
MEDSHTHCZIOVPU-UHFFFAOYSA-N
5-chloro-2,3-dihydro-1H-inden-1-one
142599
99%
42348-86-7
124°C to 125°C (3 mmHg)
MFCD00041456
1448000
Insoluble in water.
ClC1=CC=C2C(=O)CCC2=C1
166.60
166.61

Chemical Identifiers

42348-86-7
166.60
MEDSHTHCZIOVPU-UHFFFAOYSA-N
142599
ClC1=CC=C2C(=O)CCC2=C1
C9H7ClO
MFCD00041456
5-chloro-1-indanone, 5-chloro-2,3-dihydro-1h-inden-1-one, 5-chloroindan-1-one, 5-chloro-indan-1-one, 1h-inden-1-one, 5-chloro-2,3-dihydro, 5-chloroindanone, 5-chloroindone, pubchem2250, 1-indanone, 5-chloro, 5-chloro indanone
5-chloro-2,3-dihydro-1H-inden-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only