Please login to your online account to display your discounted pricing

5-Chloro-1-indanone, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Chloro-1-indanone
Name Note 98%
CAS 42348-86-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H7ClO
Formula Weight 166.61
MDL Number MFCD00041456
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC271122500

  SDS
thermo fisher scientific
271122500
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 166.61
Quantity 250mg
Color White to light yellow
Melting Point 93° to 98°C
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 5-Chloro-1-indanone
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H7ClO
MDL Number MFCD00041456
CAS 42348-86-7
CAS Min % 97.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.