missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone, TCI America™

Manufacturer:  TCI America C306825G

 View more versions of this product

Catalog No. C306825G



Specifications

Specifications

5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone
229°C
53786-28-0
MFCD02093793
DOAYWDKFDPSTSV-UHFFFAOYSA-N
6-chloro-3-piperidin-4-yl-1H-benzimidazol-2-one
104607
Crystal-Powder at 20°C
White
25g
C12H14ClN3O
5-chloro-1-4-piperidyl-2-benzimidazolinone, 5-chloro-1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 1-4-piperidinyl-1,3-dihydro-5-chlorobenzimidazolone, 5-chloro-1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one, domperidone impurity a, 5-chloro-1-piperidin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 5-chloro-1-piperidin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 5-chloro-1-piperidin-4-yl-1,3-dihydro-benzoimidazol-2-one, 4-5-chloro-2-oxo-2h-benzimidazol-1-yl piperidine, 5-chloro-1-4-piperidyl-3-hydrobenzimidazol-2-one
C1CNCCC1N2C3=C(C=C(C=C3)Cl)NC2=O
251.714
251.71
≥98.0% (GC,T)

Chemical Identifiers

53786-28-0
251.714
DOAYWDKFDPSTSV-UHFFFAOYSA-N
104607
C1CNCCC1N2C3=C(C=C(C=C3)Cl)NC2=O
C12H14ClN3O
MFCD02093793
5-chloro-1-4-piperidyl-2-benzimidazolinone, 5-chloro-1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 1-4-piperidinyl-1,3-dihydro-5-chlorobenzimidazolone, 5-chloro-1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one, domperidone impurity a, 5-chloro-1-piperidin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 5-chloro-1-piperidin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 5-chloro-1-piperidin-4-yl-1,3-dihydro-benzoimidazol-2-one, 4-5-chloro-2-oxo-2h-benzimidazol-1-yl piperidine, 5-chloro-1-4-piperidyl-3-hydrobenzimidazol-2-one
6-chloro-3-piperidin-4-yl-1H-benzimidazol-2-one
Safety and Handling

Safety and Handling

TSCA : No

Documents