missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone 98.0+%, TCI America™
$130.48 - $328.08
Chemical Identifiers
| CAS | 53786-28-0 |
|---|---|
| Molecular Formula | C12H14ClN3O |
| Molecular Weight (g/mol) | 251.714 |
| MDL Number | MFCD02093793 |
| InChI Key | DOAYWDKFDPSTSV-UHFFFAOYSA-N |
| Synonym | 5-chloro-1-4-piperidyl-2-benzimidazolinone, 5-chloro-1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 1-4-piperidinyl-1,3-dihydro-5-chlorobenzimidazolone, 5-chloro-1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one, domperidone impurity a, 5-chloro-1-piperidin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 5-chloro-1-piperidin-4-yl-1,3-dihydro-benzoimidazol-2-one, 4-5-chloro-2-oxo-2h-benzimidazol-1-yl piperidine, 5-chloro-1-4-piperidyl-3-hydrobenzimidazol-2-one |
| PubChem CID | 104607 |
| IUPAC Name | 6-chloro-3-piperidin-4-yl-1H-benzimidazol-2-one |
| SMILES | C1CNCCC1N2C3=C(C=C(C=C3)Cl)NC2=O |
Chemical Identifiers
| 53786-28-0 | |
| 251.714 | |
| DOAYWDKFDPSTSV-UHFFFAOYSA-N | |
| 104607 | |
| C1CNCCC1N2C3=C(C=C(C=C3)Cl)NC2=O |
| C12H14ClN3O | |
| MFCD02093793 | |
| 5-chloro-1-4-piperidyl-2-benzimidazolinone, 5-chloro-1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 1-4-piperidinyl-1,3-dihydro-5-chlorobenzimidazolone, 5-chloro-1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one, domperidone impurity a, 5-chloro-1-piperidin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 5-chloro-1-piperidin-4-yl-1,3-dihydro-benzoimidazol-2-one, 4-5-chloro-2-oxo-2h-benzimidazol-1-yl piperidine, 5-chloro-1-4-piperidyl-3-hydrobenzimidazol-2-one | |
| 6-chloro-3-piperidin-4-yl-1H-benzimidazol-2-one |
Specifications
| 53786-28-0 | |
| White | |
| MFCD02093793 | |
| 5-chloro-1-4-piperidyl-2-benzimidazolinone, 5-chloro-1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 1-4-piperidinyl-1,3-dihydro-5-chlorobenzimidazolone, 5-chloro-1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one, domperidone impurity a, 5-chloro-1-piperidin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 5-chloro-1-piperidin-4-yl-1,3-dihydro-benzoimidazol-2-one, 4-5-chloro-2-oxo-2h-benzimidazol-1-yl piperidine, 5-chloro-1-4-piperidyl-3-hydrobenzimidazol-2-one | |
| C1CNCCC1N2C3=C(C=C(C=C3)Cl)NC2=O | |
| 251.714 | |
| 251.71 | |
| Crystalline Powder |
| 229°C | |
| C12H14ClN3O | |
| 5 g | |
| DOAYWDKFDPSTSV-UHFFFAOYSA-N | |
| 6-chloro-3-piperidin-4-yl-1H-benzimidazol-2-one | |
| 104607 | |
| ≥98.0% (GC,T) | |
| 5-Chloro-1-(4-piperidinyl)-2-benzimidazolinone |
Safety and Handling
TSCA : No