Please login to your online account to display your discounted pricing

5-Chloro-1,3-dimethylpyrazole, 98+%, ACROS Organics™

$34.60 - $34.60

Chemical Identifiers

Chemical Name or Material 5-Chloro-1, 3-dimethylpyrazole
Name Note 98+%
CAS 54454-10-3
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C5H7ClN2
Formula Weight 130.58
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC270220010

  SDS
thermo fisher scientific
AC270220010
Glass bottle 1g Each for $34.60
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Liquid
Formula Weight 130.58
Quantity 1g
Color Clear light yellow
CAS Min % 98.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Chemical Name or Material 5-Chloro-1, 3-dimethylpyrazole
Refractive Index 1.4850 to 1.4870 (20°C, 589nm)
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C5H7ClN2
CAS 54454-10-3
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.