Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-1,3-dimethyl-4-nitro-1H-pyrazole, 97%

$105.16 - $417.38

Chemical Identifiers

Chemical Name or Material 5-Chloro-1,3-dimethyl-4-nitro-1H-pyrazole
CAS 13551-73-0
Molecular Formula C5H6ClN3O2
Formula Weight 175.58
MDL Number MFCD00052534
Beilstein 149733
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1225203

  SDS
alfa aesar™
L1225203
1g Each for $105.16

AAL1225206

  SDS
alfa aesar™
L1225206
5g Each for $417.38
Description & Specifications

Specifications

Formula Weight 175.58
Chemical Name or Material 5-Chloro-1,3-dimethyl-4-nitro-1H-pyrazole
Health Hazard 1 Warning
CAS 13551-73-0
Beilstein 149733
Assay 97%
Molecular Formula C5H6ClN3O2
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 76° to 80°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00052534
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.