missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Chloro-1,3-dimethyl-4-nitro-1H-pyrazole, 97%

Manufacturer:  Alfa Aesar™ L1225203

 View more versions of this product

Catalog No. AAL1225203



Specifications

Specifications

5-Chloro-1,3-dimethyl-4-nitro-1H-pyrazole
1g
C5H6ClN3O2
149733
WBMBDRIOUHCVAS-UHFFFAOYSA-N
5-chloro-1,3-dimethyl-4-nitropyrazole
279515
97%
76°C to 80°C
13551-73-0
MFCD00052534
5-chloro-1,3-dimethyl-4-nitro-1h-pyrazole, 1,3-dimethyl-4-nitro-5-chloropyrazole, 5-chloro-4-nitro-1,3-dimethylpyrazole, 1,3-dimethyl-4-nitro-5-chloro-1h-pyrazole, 1h-pyrazole,5-chloro-1,3-dimethyl-4-nitro, 1h-pyrazole, 5-chloro-1,3-dimethyl-4-nitro, acmc-20adw4, buttpark 3711-59, 5-chloro-1,3-dimethyl-4-nitro-1h-pyrazole #, 5-chloro-4-nitro-1,3-dimethylpyrazole,
CC1=NN(C(=C1[N+](=O)[O-])Cl)C
175.572
175.58

Chemical Identifiers

13551-73-0
175.572
WBMBDRIOUHCVAS-UHFFFAOYSA-N
279515
CC1=NN(C(=C1[N+](=O)[O-])Cl)C
C5H6ClN3O2
MFCD00052534
5-chloro-1,3-dimethyl-4-nitro-1h-pyrazole, 1,3-dimethyl-4-nitro-5-chloropyrazole, 5-chloro-4-nitro-1,3-dimethylpyrazole, 1,3-dimethyl-4-nitro-5-chloro-1h-pyrazole, 1h-pyrazole,5-chloro-1,3-dimethyl-4-nitro, 1h-pyrazole, 5-chloro-1,3-dimethyl-4-nitro, acmc-20adw4, buttpark 3711-59, 5-chloro-1,3-dimethyl-4-nitro-1h-pyrazole #, 5-chloro-4-nitro-1,3-dimethylpyrazole,
5-chloro-1,3-dimethyl-4-nitropyrazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents